Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylimidazole-5-carboxaldehyde, 97%

$166.24 - $166.24

Chemical Identifiers

Chemical Name or Material 1-Methylimidazole-5-carboxaldehyde
CAS 39021-62-0
Molecular Formula C5H6N2O
Formula Weight 110.12
MDL Number MFCD00030439
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2733803

  SDS
alfa aesar™
H2733803
1g Each for $166.24
Description & Specifications

Specifications

Formula Weight 110.12
Chemical Name or Material 1-Methylimidazole-5-carboxaldehyde
Assay 97%
Molecular Formula C5H6N2O
MDL Number MFCD00030439
CAS 39021-62-0
Health Hazard 1 Warning
Melting Point 52° to 57°C
Quantity 1g
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.