missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylimidazole-4-carboxylic acid, 95%

$72.89 - $266.85

Chemical Identifiers

CAS 41716-18-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD02179560
InChI Key WZTRQGJMMHMFGH-UHFFFAOYSA-N
Synonym 1-methyl-1h-4carboxylic acid, 1-methyl-1h-imidazole-4-carboxylic acid, 1-methyl-4-imidazole-carboxylic acid, 1-methyl-4-imidazolecarboxylic acid, 1-methyl-4-imidazolecarboxylicacid, 1-methyl-4-imidazolic acid, 1-methyl-imidazole-4-carboxylic acid, acmc-1am6x, ksc237q3j, pubchem9001
PubChem CID 541509
ChEBI CHEBI:74746
IUPAC Name 1-methylimidazole-4-carboxylic acid
SMILES CN1C=C(N=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH51866MD
View Documents
Alfa Aesar™
H51866MD
250mg
Each for $72.89
 
AAH5186603
View Documents
Alfa Aesar™
H5186603
1g
Each for $266.85
 
Description

Description

Specifications

Chemical Identifiers

41716-18-1
126.115
WZTRQGJMMHMFGH-UHFFFAOYSA-N
541509
1-methylimidazole-4-carboxylic acid
C5H6N2O2
MFCD02179560
1-methyl-1h-4carboxylic acid, 1-methyl-1h-imidazole-4-carboxylic acid, 1-methyl-4-imidazole-carboxylic acid, 1-methyl-4-imidazolecarboxylic acid, 1-methyl-4-imidazolecarboxylicacid, 1-methyl-4-imidazolic acid, 1-methyl-imidazole-4-carboxylic acid, acmc-1am6x, ksc237q3j, pubchem9001
CHEBI:74746
CN1C=C(N=C1)C(=O)O

Specifications

Hygroscopic
250mg
C5H6N2O2
1-methyl-1h-4carboxylic acid, 1-methyl-1h-imidazole-4-carboxylic acid, 1-methyl-4-imidazole-carboxylic acid, 1-methyl-4-imidazolecarboxylic acid, 1-methyl-4-imidazolecarboxylicacid, 1-methyl-4-imidazolic acid, 1-methyl-imidazole-4-carboxylic acid, acmc-1am6x, ksc237q3j, pubchem9001
CN1C=C(N=C1)C(=O)O
126.115
CHEBI:74746
95%
241°C to 243°C
41716-18-1
MFCD02179560
WZTRQGJMMHMFGH-UHFFFAOYSA-N
1-methylimidazole-4-carboxylic acid
541509
126.11
1-Methylimidazole-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS