Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylimidazole-2-carboxylic acid hydrate, 90+%

$70.82 - $227.53

Chemical Identifiers

Chemical Name or Material 1-Methylimidazole-2-carboxylic acid hydrate
CAS 20485-43-2
Molecular Formula C5H6N2O2·xH2O
Formula Weight 126.11 (Anhydrous)
MDL Number MFCD01863421
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5185903

  SDS
alfa aesar™
H5185903
1g Each for $227.53

AAH51859MD

  SDS
alfa aesar™
H51859MD
250mg Each for $70.82
Description & Specifications

Specifications

Formula Weight 126.11 (Anhydrous)
Molecular Formula C5H6N2O2·xH2O
Chemical Name or Material 1-Methylimidazole-2-carboxylic acid hydrate
Assay ≥90%
MDL Number MFCD01863421
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Melting Point 116° to 118°C
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 20485-43-2
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.