missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylhydantoin, TCI America™

$116.11 - $617.23

Chemical Identifiers

CAS 616-04-6
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.104
MDL Number MFCD00003187
InChI Key RHYBFKMFHLPQPH-UHFFFAOYSA-N
Synonym 1-methylhydantoin, dioxy-creatinine, n-methylhydantoin, hydantoin, 1-methyl, 1-methyl-hydantoin, methylhydantoin, n-methylimidazolidine-2,4-dione, unii-0546h9ur1r, 1-methyldiazolidine-2,4-dione, 2,4-imidazolidinedione, 1-methyl
PubChem CID 69217
ChEBI CHEBI:16354
IUPAC Name 1-methylimidazolidine-2,4-dione
SMILES CN1CC(=O)NC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M102725G View Documents TCI America
M102725G
25g Each for $116.11
 
M1027250G View Documents TCI America
M1027250G
250g Each for $617.23
 
Specifications

Chemical Identifiers

616-04-6
114.104
RHYBFKMFHLPQPH-UHFFFAOYSA-N
69217
1-methylimidazolidine-2,4-dione
C4H6N2O2
MFCD00003187
1-methylhydantoin, dioxy-creatinine, n-methylhydantoin, hydantoin, 1-methyl, 1-methyl-hydantoin, methylhydantoin, n-methylimidazolidine-2,4-dione, unii-0546h9ur1r, 1-methyldiazolidine-2,4-dione, 2,4-imidazolidinedione, 1-methyl
CHEBI:16354
CN1CC(=O)NC1=O

Specifications

White
25g
C4H6N2O2
1-methylhydantoin, dioxy-creatinine, n-methylhydantoin, hydantoin, 1-methyl, 1-methyl-hydantoin, methylhydantoin, n-methylimidazolidine-2,4-dione, unii-0546h9ur1r, 1-methyldiazolidine-2,4-dione, 2,4-imidazolidinedione, 1-methyl
CN1CC(=O)NC1=O
114.104
CHEBI:16354
Crystal-Powder at 20°C
1-Methylhydantoin
158°C
616-04-6
MFCD00003187
RHYBFKMFHLPQPH-UHFFFAOYSA-N
1-methylimidazolidine-2,4-dione
69217
114.10
≥99.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6017

TSCA : No