missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylhydantoin 99.0+%, TCI America™

Manufacturer:  Tci America M1027250G

 View more versions of this product

Catalog No. M1027250G



Specifications

Specifications

1-Methylhydantoin
158°C
616-04-6
MFCD00003187
RHYBFKMFHLPQPH-UHFFFAOYSA-N
1-methylimidazolidine-2,4-dione
69217
114.10
≥99.0% (T)
White
250g
C4H6N2O2
1-methylhydantoin, dioxy-creatinine, n-methylhydantoin, hydantoin, 1-methyl, 1-methyl-hydantoin, methylhydantoin, n-methylimidazolidine-2,4-dione, unii-0546h9ur1r, 1-methyldiazolidine-2,4-dione, 2,4-imidazolidinedione, 1-methyl
CN1CC(=O)NC1=O
114.104
CHEBI:16354
Crystal-Powder at 20°C

Chemical Identifiers

616-04-6
114.104
RHYBFKMFHLPQPH-UHFFFAOYSA-N
69217
1-methylimidazolidine-2,4-dione
C4H6N2O2
MFCD00003187
1-methylhydantoin, dioxy-creatinine, n-methylhydantoin, hydantoin, 1-methyl, 1-methyl-hydantoin, methylhydantoin, n-methylimidazolidine-2,4-dione, unii-0546h9ur1r, 1-methyldiazolidine-2,4-dione, 2,4-imidazolidinedione, 1-methyl
CHEBI:16354
CN1CC(=O)NC1=O
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6017

TSCA : No

Documents