Please login to your online account to display your discounted pricing

1-Methylhydantoin 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Methylhydantoin
Name Note 97%
CAS 616-04-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H6N2O2
Formula Weight 114.1
MDL Number MFCD00003187
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC126940050

  SDS
fisher scientific acros
126940050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystalline Powder
Assay ≥96.0%
Formula Weight 114.1
Quantity 5g
Color White
EINECS Number 210-460-6
TSCA TSCA
Melting Point 156° to 160°C
Beilstein 24, 244
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 1-Methylhydantoin
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C4H6N2O2
MDL Number MFCD00003187
CAS 616-04-6