Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylhydantoin, 97%

$41.58 - $109.09

Chemical Identifiers

Chemical Name or Material 1-Methylhydantoin
CAS 616-04-6
Molecular Formula C4H6N2O2
Formula Weight 114.1
MDL Number MFCD00003187
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2432206

  SDS
alfa aesar™
B2432206
5g Each for $41.58

AAB2432214

  SDS
alfa aesar™
B2432214
25g Each for $109.09
Description & Specifications

Specifications

Formula Weight 114.1
Melting Point 156° to 159°C
Assay 97%
Molecular Formula C4H6N2O2
Chemical Name or Material 1-Methylhydantoin
MDL Number MFCD00003187
CAS 616-04-6
Quantity 5g

Applications
1-Methylhydantoin is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. 1-Methylhydantoin and 5-hydroxy-1-methylhydantoin were isolated as plant growth regulators from rabbit skin tissue which had been inflamed by inoculation with vaccinia virus.

Solubility
Soluble in methanol. (almost transparency). Insoluble in water.

Notes
Store at 4°C. Incompatible with oxidizing agents.