missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylhomopiperazine, TCI America™

Manufacturer:  TCI America M075125ML

 View more versions of this product

Catalog No. M075125ML



Specifications

Specifications

1-Methylhomopiperazine
Yellow
4318-37-0
MFCD00059810
n-methylhomopiperazine, n-methyl homopiperazine, 1-methylhomopiperazine, 1h-1,4-diazepine, hexahydro-1-methyl, hexahydro-1-methyl-1h-1,4-diazepine, 1-methyl-1,4-diazaperhydroepine, methylhomopiperazine, n-methylhomopiperizine, 4-methylhomopiperazine, 1-methylhomopiperizine
CN1CCCNCC1
114.192
114.19
≥98.0% (GC,T)
164°C
25mL
C6H14N2
2920
FXHRAKUEZPSMLJ-UHFFFAOYSA-N
1-methyl-1,4-diazepane
228349
Clear Liquid at 20°C

Chemical Identifiers

4318-37-0
114.192
FXHRAKUEZPSMLJ-UHFFFAOYSA-N
228349
CN1CCCNCC1
C6H14N2
MFCD00059810
n-methylhomopiperazine, n-methyl homopiperazine, 1-methylhomopiperazine, 1h-1,4-diazepine, hexahydro-1-methyl, hexahydro-1-methyl-1h-1,4-diazepine, 1-methyl-1,4-diazaperhydroepine, methylhomopiperazine, n-methylhomopiperizine, 4-methylhomopiperazine, 1-methylhomopiperizine
1-methyl-1,4-diazepane
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1386

TSCA : No

Documents