missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylhomopiperazine, TCI America™

$82.50 - $267.36

Chemical Identifiers

CAS 4318-37-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00059810
InChI Key FXHRAKUEZPSMLJ-UHFFFAOYSA-N
Synonym n-methylhomopiperazine, n-methyl homopiperazine, 1-methylhomopiperazine, 1h-1,4-diazepine, hexahydro-1-methyl, hexahydro-1-methyl-1h-1,4-diazepine, 1-methyl-1,4-diazaperhydroepine, methylhomopiperazine, n-methylhomopiperizine, 4-methylhomopiperazine, 1-methylhomopiperizine
PubChem CID 228349
IUPAC Name 1-methyl-1,4-diazepane
SMILES CN1CCCNCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M075125ML View Documents TCI America
M075125ML
25mL Each for $267.36
 
M07515ML View Documents TCI America
M07515ML
5mL Each for $82.50
 
Specifications

Chemical Identifiers

4318-37-0
114.192
FXHRAKUEZPSMLJ-UHFFFAOYSA-N
228349
CN1CCCNCC1
C6H14N2
MFCD00059810
n-methylhomopiperazine, n-methyl homopiperazine, 1-methylhomopiperazine, 1h-1,4-diazepine, hexahydro-1-methyl, hexahydro-1-methyl-1h-1,4-diazepine, 1-methyl-1,4-diazaperhydroepine, methylhomopiperazine, n-methylhomopiperizine, 4-methylhomopiperazine, 1-methylhomopiperizine
1-methyl-1,4-diazepane

Specifications

164°C
25mL
C6H14N2
2920
FXHRAKUEZPSMLJ-UHFFFAOYSA-N
1-methyl-1,4-diazepane
228349
Clear Liquid at 20°C
1-Methylhomopiperazine
Yellow
4318-37-0
MFCD00059810
n-methylhomopiperazine, n-methyl homopiperazine, 1-methylhomopiperazine, 1h-1,4-diazepine, hexahydro-1-methyl, hexahydro-1-methyl-1h-1,4-diazepine, 1-methyl-1,4-diazaperhydroepine, methylhomopiperazine, n-methylhomopiperizine, 4-methylhomopiperazine, 1-methylhomopiperizine
CN1CCCNCC1
114.192
114.19
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1386

TSCA : No