missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylhomopiperazine, 97%

Manufacturer:  Alfa Aesar™ H55441AE

 View more versions of this product

Catalog No. AAH55441AE



Specifications

Specifications

1-Methylhomopiperazine
74°C to 75°C (35mmHg)
4318-37-0
MFCD00059810
1-methyl-1,4-diazaperhydroepine
CN1CCCNCC1
114.192
114.19
45°C (113°F)
100mL
C6H14N2
UN2920
FXHRAKUEZPSMLJ-UHFFFAOYSA-N
1-methyl-1,4-diazepane
228349
97%

Chemical Identifiers

4318-37-0
114.192
FXHRAKUEZPSMLJ-UHFFFAOYSA-N
228349
CN1CCCNCC1
C6H14N2
MFCD00059810
1-methyl-1,4-diazaperhydroepine
1-methyl-1,4-diazepane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

Documents