missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylhomopiperazine, 97%

Manufacturer:  Alfa Aesar™ H55441AC

 View more versions of this product

Catalog No. AAH55441AC



Description

Description

Specifications

Specifications

1-Methylhomopiperazine
45°C (113°F)
74°C to 75°C (35mmHg)
4318-37-0
MFCD00059810
1-methyl-1,4-diazaperhydroepine, 1-methylhomopiperazine, 1-methylhomopiperizine, 1h-1,4-diazepine, hexahydro-1-methyl, 4-methylhomopiperazine, hexahydro-1-methyl-1h-1,4-diazepine, methylhomopiperazine, n-methyl homopiperazine, n-methylhomopiperazine, n-methylhomopiperizine
CN1CCCNCC1
114.192
114.19
0.918
1.477
25mL
C6H14N2
UN2920
FXHRAKUEZPSMLJ-UHFFFAOYSA-N
1-methyl-1,4-diazepane
228349
97%

Chemical Identifiers

4318-37-0
114.192
FXHRAKUEZPSMLJ-UHFFFAOYSA-N
228349
CN1CCCNCC1
C6H14N2
MFCD00059810
1-methyl-1,4-diazaperhydroepine, 1-methylhomopiperazine, 1-methylhomopiperizine, 1h-1,4-diazepine, hexahydro-1-methyl, 4-methylhomopiperazine, hexahydro-1-methyl-1h-1,4-diazepine, methylhomopiperazine, n-methyl homopiperazine, n-methylhomopiperazine, n-methylhomopiperizine
1-methyl-1,4-diazepane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents