missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylguanidine Hydrochloride 98.0+%, TCI America™

Manufacturer:  TCI America M033625G

 View more versions of this product

Catalog No. M033625G



Specifications

Specifications

1-Methylguanidine Hydrochloride
25g
C2H8ClN3
VJQCNCOGZPSOQZ-UHFFFAOYSA-N
2-methylguanidine;hydrochloride
146724
Crystal-Powder at 20°C
White
21770-81-0
MFCD00012576
CN=C(N)N.Cl
109.557
109.56
≥98.0% (N,T)

Chemical Identifiers

21770-81-0
109.557
VJQCNCOGZPSOQZ-UHFFFAOYSA-N
2-methylguanidine;hydrochloride
C2H8ClN3
MFCD00012576
146724
CN=C(N)N.Cl
Safety and Handling

Safety and Handling

RTECSNumber : MF3990000

TSCA : No

Documents