missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylcyclopentene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1881606

 View more versions of this product

Catalog No. AAA1881606



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Methylcyclopentene
-142°C
73°C to 75°C
C6H10
MFCD00001397
UN3295
1-methyl-1-cyclopentene, cyclopentene, 1-methyl, methylcyclopentene, cyclopentene, methyl, unii-v03m1dzc9i, 1-methylcyclopent-1-ene, v03m1dzc9i, methyl-cyclopentene, 1-methylcyclopentene-1, cyc1opentene,l-methyl
CC1=CCCC1
82.15
82.15
693-89-0
0.779
−17°C (1°F)
1.431
5 g
1900640
ATQUFXWBVZUTKO-UHFFFAOYSA-N
1-methylcyclopentene
12746
98%

Chemical Identifiers

693-89-0
82.15
ATQUFXWBVZUTKO-UHFFFAOYSA-N
12746
CC1=CCCC1
C6H10
MFCD00001397
1-methyl-1-cyclopentene, cyclopentene, 1-methyl, methylcyclopentene, cyclopentene, methyl, unii-v03m1dzc9i, 1-methylcyclopent-1-ene, v03m1dzc9i, methyl-cyclopentene, 1-methylcyclopentene-1, cyc1opentene,l-methyl
1-methylcyclopentene
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H225-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 211-762-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only