Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylcyclohexanol, 97%

Chemical Identifiers

Chemical Name or Material 1-Methylcyclohexanol
CAS 590-67-0
Molecular Formula C7H14O
Formula Weight 114.19
MDL Number MFCD00003857
UN Number UN2617
Beilstein 1900934
Synonym Cyclohexyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0096706

 
alfa aesar™
L0096706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.19
Chemical Name or Material 1-Methylcyclohexanol
CAS 590-67-0
Assay 97%
Molecular Formula C7H14O
MDL Number MFCD00003857
UN Number UN2617
Beilstein 1900934
Synonym Cyclohexyl methyl carbinol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H332
Flammable solid.
Harmful if inhaled.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYLCYCLOHEXANOLS
Flash Point 55°C (131°F)
Boiling Point 168° to 169°C
Melting Point 25° to 27°C
Quantity 5g
Health Hazard 3 GHS P Statement
P210-P261-P280-P240-P241-P304+P340
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P240-P241-P304+P340
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS H Statement
H228-H332
Flammable solid.
Harmful if inhaled.