Learn More
1-Methylcyclohexanol, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 126601000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Methylcyclohexanol | |
95.0 | |
0.9190g/mL | |
67°C | |
95% min. (GC) | |
C7H14O | |
CH3C6H10OH | |
100g | |
0.919 | |
VTBOTOBFGSVRMA-UHFFFAOYSA-N | |
1-methylcyclohexan-1-ol | |
11550 | |
96% |
590-67-0 | |
100.0 | |
168°C (752.0mmHg) | |
Authentic | |
Glass bottle | |
1.4590 to 1.461 | |
MFCD00003857 | |
06, 11 | |
1-methylcyclohexanol, cyclohexanol, 1-methyl, 1-methyl-1-cyclohexanol, cyclohexanol,methyl, 1-methyl-cyclohexanol, 1-methylcyclohexyl alcohol, acmc-1b1ai, 1-methyl-cyclohexan-1-ol, 1-methyl cyclohexanol | |
CC1(CCCCC1)O | |
114.19 | |
114.19 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Flammable solid.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/va
GHS Signal Word: Warning
EINECSNumber : 209-688-9
RUO â Research Use Only