Learn More
1-Methylcyclohexanol, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 126600250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Methylcyclohexanol | |
95.0 | |
Colorless to Yellow | |
Authentic | |
Glass bottle | |
1.4590 to 1.4610 (20°C, 589nm) | |
MFCD00003857 | |
06, 11 | |
VTBOTOBFGSVRMA-UHFFFAOYSA-N | |
1-methylcyclohexan-1-ol | |
11550 | |
≥95% |
590-67-0 | |
100.0 | |
0.9190g/mL | |
96% | |
C7H14O | |
CH3C6H10OH | |
25 g | |
1-methylcyclohexanol, cyclohexanol, 1-methyl, 1-methyl-1-cyclohexanol, cyclohexanol,methyl, 1-methyl-cyclohexanol, 1-methylcyclohexyl alcohol, acmc-1b1ai, 1-methyl-cyclohexan-1-ol, 1-methyl cyclohexanol | |
CC1(CCCCC1)O | |
114.188 | |
114.19 | |
Liquid After Melting |
Chemical Identifiers
590-67-0 | |
VTBOTOBFGSVRMA-UHFFFAOYSA-N | |
11550 | |
CC1(CCCCC1)O |
MFCD00003857 | |
1-methylcyclohexanol, cyclohexanol, 1-methyl, 1-methyl-1-cyclohexanol, cyclohexanol,methyl, 1-methyl-cyclohexanol, 1-methylcyclohexyl alcohol, acmc-1b1ai, 1-methyl-cyclohexan-1-ol, 1-methyl cyclohexanol | |
1-methylcyclohexan-1-ol |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only