Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methylbenzimidazole-2-carboxaldehyde, 98+%

$204.73 - $831.22

Chemical Identifiers

Chemical Name or Material 1-Methylbenzimidazole-2-carboxaldehyde
CAS 3012-80-4
Molecular Formula C9H8N2O
Formula Weight 160.17
MDL Number MFCD00142655
Synonym 1-Methyl-2-formylbenzimidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2560103

 
alfa aesar™
B2560103
1g Each for $204.73

AAB2560106

 
alfa aesar™
B2560106
5g Each for $831.22
Description & Specifications

Specifications

Formula Weight 160.17
Chemical Name or Material 1-Methylbenzimidazole-2-carboxaldehyde
Assay ≥98%
Molecular Formula C9H8N2O
MDL Number MFCD00142655
CAS 3012-80-4
Synonym 1-Methyl-2-formylbenzimidazole
Health Hazard 1 Warning
Melting Point 120° to 122°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.