missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(Methylamino)anthraquinone 98.0+%, TCI America™
$97.89 - $744.89
Chemical Identifiers
CAS | 82-38-2 |
---|---|
Molecular Formula | C15H11NO2 |
Molecular Weight (g/mol) | 237.258 |
MDL Number | MFCD00001197 |
InChI Key | SVTDYSXXLJYUTM-UHFFFAOYSA-N |
PubChem CID | 6706 |
IUPAC Name | 1-(methylamino)anthracene-9,10-dione |
SMILES | CNC1=CC=CC2=C1C(=O)C3=CC=CC=C3C2=O |
Chemical Identifiers
82-38-2 | |
237.258 | |
SVTDYSXXLJYUTM-UHFFFAOYSA-N | |
1-(methylamino)anthracene-9,10-dione |
C15H11NO2 | |
MFCD00001197 | |
6706 | |
CNC1=CC=CC2=C1C(=O)C3=CC=CC=C3C2=O |
Specifications
82-38-2 | |
C15H11NO2 | |
25 g | |
CNC1=CC=CC2=C1C(=O)C3=CC=CC=C3C2=O | |
237.258 | |
237.26 | |
Crystalline Powder |
169°C | |
MFCD00001197 | |
SVTDYSXXLJYUTM-UHFFFAOYSA-N | |
1-(methylamino)anthracene-9,10-dione | |
6706 | |
≥98.0% (HPLC) | |
1-(Methylamino)anthraquinone |
Safety and Handling
EINECSNumber : (4)-0708
RTECSNumber : CB0536600
TSCA : Yes