missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl L-Glutamate, TCI America™

$151.25 - $611.13

Chemical Identifiers

CAS 6384-08-3
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
MDL Number MFCD00171677
InChI Key SEWIYICDCVPBEW-UHFFFAOYNA-N
Synonym h-glu-ome, s-4-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid 1-methyl ester, l-glutamate methylester, 1-methyl l-glutamate, l-glutamate methyl ester, glutamic acid methyl ester, 4s-4-amino-5-methoxy-5-oxopentanoic acid, gluome, glu alpha-ome-oh
PubChem CID 440550
ChEBI CHEBI:28575
IUPAC Name (4S)-4-amino-5-methoxy-5-oxopentanoic acid
SMILES COC(=O)C(CCC(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M186125G View Documents TCI America
M186125G
25g Each for $611.13
 
M18615G View Documents TCI America
M18615G
5g Each for $151.25
 
Specifications

Chemical Identifiers

6384-08-3
161.16
SEWIYICDCVPBEW-UHFFFAOYNA-N
440550
(4S)-4-amino-5-methoxy-5-oxopentanoic acid
C6H11NO4
MFCD00171677
h-glu-ome, s-4-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid 1-methyl ester, l-glutamate methylester, 1-methyl l-glutamate, l-glutamate methyl ester, glutamic acid methyl ester, 4s-4-amino-5-methoxy-5-oxopentanoic acid, gluome, glu alpha-ome-oh
CHEBI:28575
COC(=O)C(CCC(=O)O)N

Specifications

White
25g
C6H11NO4
h-glu-ome, s-4-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid 1-methyl ester, l-glutamate methylester, 1-methyl l-glutamate, l-glutamate methyl ester, glutamic acid methyl ester, 4s-4-amino-5-methoxy-5-oxopentanoic acid, gluome, glu alpha-ome-oh
COC(=O)C(CCC(=O)O)N
161.16
CHEBI:28575
Crystal-Powder at 20°C
1-Methyl L-Glutamate
145°C
6384-08-3
MFCD00171677
SEWIYICDCVPBEW-UHFFFAOYNA-N
(4S)-4-amino-5-methoxy-5-oxopentanoic acid
440550
161.16
≥98.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0418

TSCA : No