Please login to your online account to display your discounted pricing

1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  BTB09194DE

 View more versions of this product

Catalog No. BTB09194DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 166.1
Quantity 5g
Assay 97%
CAS Max % 100.0
CAS 119022-51-4
Chemical Name or Material 1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol
Molecular Formula C5H5F3N2O
CAS Min % 96.5
MDL Number MFCD00085094
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol
CAS 119022-51-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5F3N2O
Formula Weight 166.1
MDL Number MFCD00085094