Please login to your online account to display your discounted pricing

1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol
CAS 119022-51-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5F3N2O
Formula Weight 166.1
MDL Number MFCD00085094
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB09194DE

 
fisher scientific company
BTB09194DE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB09194DA

  SDS
fisher scientific company
BTB09194DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.1
Quantity 5g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol
Assay Percent Range 97%
Molecular Formula C5H5F3N2O
MDL Number MFCD00085094
CAS 119022-51-4
CAS Min % 96.5