Please login to your online account to display your discounted pricing

1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC29541CB

 View more versions of this product

Catalog No. CC29541CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 439.15
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material 1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole
Molecular Formula C17H31F3N2Sn
MDL Number MFCD06797466
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
CAS Min % 96.5
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 191606-78-7
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole
CAS 191606-78-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C17H31F3N2Sn
Formula Weight 439.15
MDL Number MFCD06797466
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.

SDS & Safety Documents