Please login to your online account to display your discounted pricing

1-Methyl-5-tributylstannanyl-1H-imidazole, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC23641DE

 View more versions of this product

Catalog No. CC23641DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 371.15
Quantity 5g
Assay 97%
CAS 147716-03-8
Chemical Name or Material 1-Methyl-5-tributylstannanyl-1H-imidazole
Molecular Formula C16H32N2Sn
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-Methyl-5-tributylstannanyl-1H-imidazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H32N2Sn
Formula Weight 371.15