Please login to your online account to display your discounted pricing

1-Methyl-5-tributylstannanyl-1H-imidazole, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Methyl-5-tributylstannanyl-1H-imidazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H32N2Sn
Formula Weight 371.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC23641DE

  SDS
fisher scientific company
CC23641DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC23641CB

  SDS
fisher scientific company
CC23641CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 371.15
Quantity 5g
Assay 97%
CAS 147716-03-8
Chemical Name or Material 1-Methyl-5-tributylstannanyl-1H-imidazole
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C16H32N2Sn