Please login to your online account to display your discounted pricing

1-Methyl-5-tributylstannanyl-1H-imidazole, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC23641CB

 View more versions of this product

Catalog No. CC23641CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 371.15
Quantity 250mg
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H32N2Sn
Chemical Name or Material 1-Methyl-5-tributylstannanyl-1H-imidazole
CAS 147716-03-8

Specifications

Chemical Name or Material 1-Methyl-5-tributylstannanyl-1H-imidazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H32N2Sn
Formula Weight 371.15