Please login to your online account to display your discounted pricing

1-Methyl-5-phenyl-1H-pyrazole-4-carbaldehyde, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Methyl-5-phenyl-1H-pyrazole-4-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H10N2O
Formula Weight 186.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC28104DE

 
fisher scientific company
CC28104DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC28104DA

 
fisher scientific company
CC28104DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC28104CB

 
fisher scientific company
CC28104CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.21
Quantity 5g
Assay 97%
Chemical Name or Material 1-Methyl-5-phenyl-1H-pyrazole-4-carbaldehyde
Assay Percent Range 97%
CAS 154927-01-2
Packaging Amber Glass Bottle
Molecular Formula C11H10N2O