missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Methyl-5-phenyl-1H-pyrazole-4-carbaldehyde, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC28104CB
Specifications
1-Methyl-5-phenyl-1H-pyrazole-4-carbaldehyde | |
97% | |
C11H10N2O | |
1-methyl-5-phenyl-1h-pyrazole-4-carbaldehyde, 1-methyl-5-phenyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde,1-methyl-5-phenyl | |
CN1C(=C(C=N1)C=O)C2=CC=CC=C2 | |
186.214 | |
186.21 |
154927-01-2 | |
Amber Glass Bottle | |
250mg | |
DLMVCAXLQMJYHP-UHFFFAOYSA-N | |
1-methyl-5-phenylpyrazole-4-carbaldehyde | |
2795475 | |
97% |
Chemical Identifiers
154927-01-2 | |
186.214 | |
1-methyl-5-phenyl-1h-pyrazole-4-carbaldehyde, 1-methyl-5-phenyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde,1-methyl-5-phenyl | |
1-methyl-5-phenylpyrazole-4-carbaldehyde |
C11H10N2O | |
DLMVCAXLQMJYHP-UHFFFAOYSA-N | |
2795475 | |
CN1C(=C(C=N1)C=O)C2=CC=CC=C2 |