Please login to your online account to display your discounted pricing

1-Methyl-5-phenyl-1H-pyrazole-4-carbaldehyde, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC28104DA

 View more versions of this product

Catalog No. CC28104DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 186.21
Quantity 1g
Assay 97%
Molecular Formula C11H10N2O
Assay Percent Range 97%
CAS 154927-01-2
Chemical Name or Material 1-Methyl-5-phenyl-1H-pyrazole-4-carbaldehyde
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 1-Methyl-5-phenyl-1H-pyrazole-4-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H10N2O
Formula Weight 186.21