missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-4-piperidone, TCI America™

$36.67 - $291.81

Chemical Identifiers

CAS 1445-73-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006191
InChI Key HUUPVABNAQUEJW-UHFFFAOYSA-N
Synonym 1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one
PubChem CID 74049
IUPAC Name 1-methylpiperidin-4-one
SMILES CN1CCC(=O)CC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M053025G View Documents TCI America
M053025G
25g Each for $36.67
 
M0530500G View Documents TCI America
M0530500G
500g Each for $291.81
 
M0530100G View Documents TCI America
M0530100G
100g Each for $96.25
 
Specifications

Chemical Identifiers

1445-73-4
113.16
HUUPVABNAQUEJW-UHFFFAOYSA-N
74049
CN1CCC(=O)CC1
C6H11NO
MFCD00006191
1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one
1-methylpiperidin-4-one

Specifications

72°C
25g
C6H11NO
1224
HUUPVABNAQUEJW-UHFFFAOYSA-N
1-methylpiperidin-4-one
74049
Clear Liquid at 20°C
1-Methyl-4-piperidone
Yellow
1445-73-4
MFCD00006191
1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one
CN1CCC(=O)CC1
113.16
113.16
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-2198

TSCA : Yes