missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Methyl-4-piperidone 98.0+%, TCI America™
Supplier: TCI America M0530500G
Specifications
1-Methyl-4-piperidone | |
Yellow | |
C6H11NO | |
500 g | |
1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one | |
CN1CCC(=O)CC1 | |
113.16 | |
113.16 | |
Liquid |
1445-73-4 | |
72°C | |
MFCD00006191 | |
1224 | |
HUUPVABNAQUEJW-UHFFFAOYSA-N | |
1-methylpiperidin-4-one | |
74049 | |
≥98.0% (GC,T) |
Chemical Identifiers
1445-73-4 | |
113.16 | |
HUUPVABNAQUEJW-UHFFFAOYSA-N | |
74049 | |
CN1CCC(=O)CC1 |
C6H11NO | |
MFCD00006191 | |
1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one | |
1-methylpiperidin-4-one |
Safety and Handling
EINECSNumber : (9)-2198
TSCA : Yes