Learn More
1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
$131.82 - $577.65
Chemical Identifiers
CAS | 1445-73-4 |
---|---|
Molecular Formula | C6H11NO |
Molecular Weight (g/mol) | 113.16 |
MDL Number | MFCD00006191 |
InChI Key | HUUPVABNAQUEJW-UHFFFAOYSA-N |
Synonym | 1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one |
PubChem CID | 74049 |
IUPAC Name | 1-methylpiperidin-4-one |
SMILES | CN1CCC(=O)CC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC156921000
|
Thermo Scientific Chemicals
156921000 |
100 g | Glass bottle |
Each for $131.82
|
|
||||
AC156925000
|
Thermo Scientific Chemicals
156925000 |
500 g | Glass bottle |
Each for $577.65
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1445-73-4 | |
113.16 | |
HUUPVABNAQUEJW-UHFFFAOYSA-N | |
74049 | |
CN1CCC(=O)CC1 |
C6H11NO | |
MFCD00006191 | |
1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one | |
1-methylpiperidin-4-one |
Specifications
1445-73-4 | |
100.0 | |
0.9800g/mL | |
58°C | |
97.5% min. (GC) | |
C6H11NO | |
MFCD00006191 | |
21, II, 215 | |
1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one | |
HUUPVABNAQUEJW-UHFFFAOYSA-N | |
1-methylpiperidin-4-one | |
74049 | |
98% | |
1-Methyl-4-piperidone, 98% |
97.5 | |
Yellow to Orange | |
55.0°C to 58.0°C (10.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4600 to 1.4620 | |
100 g | |
0.98 | |
Solubility in water: miscible with water. | |
CN1CCC(=O)CC1 | |
113.16 | |
113.16 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 215-895-5
TSCA : TSCA
RUO – Research Use Only