Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-4-piperidone, 98%

Chemical Identifiers

Chemical Name or Material 1-Methyl-4-piperidone
CAS 1445-73-4
Molecular Formula C6H11NO
Formula Weight 113.16
MDL Number MFCD00006191
UN Number UN2733
Beilstein 106924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA133520B

  SDS
alfa aesar™
A133520B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1335218

  SDS
alfa aesar™
A1335218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1335222

  SDS
alfa aesar™
A1335222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1335230

  SDS
alfa aesar™
A1335230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 113.16
Odor Amine-like
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Health Hazard 1 Danger
UN Number UN2733
Quantity 1000g
Assay 98%
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Boiling Point 55° to 58°C (10mmHg)
Beilstein 106924
Chemical Name or Material 1-Methyl-4-piperidone
MDL Number MFCD00006191
CAS 1445-73-4
Flash Point 60°C (140°F)
pH 12
Molecular Formula C6H11NO
Recommended Storage 2° to 8°C

Applications
1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.

Solubility
It is miscible with water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong acids.

Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

2° to 8°C