missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-4-piperidinemethanol, TCI America™

$113.06 - $366.67

Chemical Identifiers

CAS 20691-89-8
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD03411903
InChI Key KJZLJGZZDNGGCA-UHFFFAOYSA-N
Synonym 1-methyl-4-piperidinemethanol, 1-methylpiperidin-4-yl methanol, n-methyl-4-piperidinemethanol, 4-piperidinemethanol, 1-methyl, 1-methyl-4-piperidinyl methanol, 4-hydroxymethyl-1-methylpiperidine, 1-methylpiperidine-4-methanol, n-methyl-4-hydroxymethylpiperidine, 1-methyl-piperidin-4-yl-methanol, 1-methyl-4-piperidyl methan-1-ol
PubChem CID 271971
IUPAC Name (1-methylpiperidin-4-yl)methanol
SMILES CN1CCC(CC1)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M22425G View Documents TCI America
M22425G
5g Each for $113.06
 
M224225G View Documents TCI America
M224225G
25g Each for $366.67
 
Specifications

Chemical Identifiers

20691-89-8
129.203
KJZLJGZZDNGGCA-UHFFFAOYSA-N
271971
CN1CCC(CC1)CO
C7H15NO
MFCD03411903
1-methyl-4-piperidinemethanol, 1-methylpiperidin-4-yl methanol, n-methyl-4-piperidinemethanol, 4-piperidinemethanol, 1-methyl, 1-methyl-4-piperidinyl methanol, 4-hydroxymethyl-1-methylpiperidine, 1-methylpiperidine-4-methanol, n-methyl-4-hydroxymethylpiperidine, 1-methyl-piperidin-4-yl-methanol, 1-methyl-4-piperidyl methan-1-ol
(1-methylpiperidin-4-yl)methanol

Specifications

108°C
5g
C7H15NO
1-methyl-4-piperidinemethanol, 1-methylpiperidin-4-yl methanol, n-methyl-4-piperidinemethanol, 4-piperidinemethanol, 1-methyl, 1-methyl-4-piperidinyl methanol, 4-hydroxymethyl-1-methylpiperidine, 1-methylpiperidine-4-methanol, n-methyl-4-hydroxymethylpiperidine, 1-methyl-piperidin-4-yl-methanol, 1-methyl-4-piperidyl methan-1-ol
CN1CCC(CC1)CO
129.203
129.20
≥97.0% (GC,T)
Yellow
20691-89-8
MFCD03411903
KJZLJGZZDNGGCA-UHFFFAOYSA-N
(1-methylpiperidin-4-yl)methanol
271971
Clear Liquid at 20°C
1-Methyl-4-piperidinemethanol
Safety and Handling

Safety and Handling

TSCA : No