missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-4-(methylamino)piperidine, TCI America™

$27.50 - $84.03

Chemical Identifiers

CAS 73579-08-5
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00006498
InChI Key XRYGCVVVDCEPRL-UHFFFAOYSA-N
PubChem CID 566323
IUPAC Name N,1-dimethylpiperidin-4-amine
SMILES CNC1CCN(CC1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25661G View Documents TCI America
M25661G
1g Each for $84.03
 
M2566200MG View Documents TCI America
M2566200MG
200mg Each for $27.50
 
Specifications

Chemical Identifiers

73579-08-5
128.219
XRYGCVVVDCEPRL-UHFFFAOYSA-N
N,1-dimethylpiperidin-4-amine
C7H16N2
MFCD00006498
566323
CNC1CCN(CC1)C

Specifications

53°C
1g
C7H16N2
2920
CNC1CCN(CC1)C
128.219
128.22
≥97.0% (GC,T)
Undesignated
73579-08-5
MFCD00006498
XRYGCVVVDCEPRL-UHFFFAOYSA-N
N,1-dimethylpiperidin-4-amine
566323
Clear Liquid at 20°C
1-Methyl-4-(methylamino)piperidine
Safety and Handling

Safety and Handling

TSCA : No