missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-4-imidazolecarboxylic Acid 98.0+%, TCI America™

$88.88 - $263.34

Chemical Identifiers

CAS 41716-18-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD02179560
InChI Key WZTRQGJMMHMFGH-UHFFFAOYSA-N
Synonym 1-methyl-1h-4carboxylic acid, 1-methyl-1h-imidazole-4-carboxylic acid, 1-methyl-4-imidazole-carboxylic acid, 1-methyl-4-imidazolecarboxylic acid, 1-methyl-4-imidazolecarboxylicacid, 1-methyl-4-imidazolic acid, 1-methyl-imidazole-4-carboxylic acid, acmc-1am6x, ksc237q3j, pubchem9001
PubChem CID 541509
ChEBI CHEBI:74746
IUPAC Name 1-methylimidazole-4-carboxylic acid
SMILES CN1C=C(N=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M22721G View Documents Tci America
M22721G
1g Each for $88.88
 
M22725G View Documents Tci America
M22725G
5g Each for $263.34
 
Specifications

Chemical Identifiers

41716-18-1
126.115
WZTRQGJMMHMFGH-UHFFFAOYSA-N
541509
1-methylimidazole-4-carboxylic acid
C5H6N2O2
MFCD02179560
1-methyl-1h-4carboxylic acid, 1-methyl-1h-imidazole-4-carboxylic acid, 1-methyl-4-imidazole-carboxylic acid, 1-methyl-4-imidazolecarboxylic acid, 1-methyl-4-imidazolecarboxylicacid, 1-methyl-4-imidazolic acid, 1-methyl-imidazole-4-carboxylic acid, acmc-1am6x, ksc237q3j, pubchem9001
CHEBI:74746
CN1C=C(N=C1)C(=O)O

Specifications

Undesignated
1g
C5H6N2O2
1-methyl-1h-4carboxylic acid, 1-methyl-1h-imidazole-4-carboxylic acid, 1-methyl-4-imidazole-carboxylic acid, 1-methyl-4-imidazolecarboxylic acid, 1-methyl-4-imidazolecarboxylicacid, 1-methyl-4-imidazolic acid, 1-methyl-imidazole-4-carboxylic acid, acmc-1am6x, ksc237q3j, pubchem9001
CN1C=C(N=C1)C(=O)O
126.115
CHEBI:74746
Crystal-Powder at 20°C
1-Methyl-4-imidazolecarboxylic Acid
247°C
41716-18-1
MFCD02179560
WZTRQGJMMHMFGH-UHFFFAOYSA-N
1-methylimidazole-4-carboxylic acid
541509
126.12
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No