missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-4-(4-nitrophenyl)piperazine, 97%

$28.84 - $337.73

Chemical Identifiers

CAS 16155-03-6
Molecular Formula C11H15N3O2
Molecular Weight (g/mol) 221.26
MDL Number MFCD00101003
InChI Key GZNDUKANJZIZOT-UHFFFAOYSA-N
Synonym 1-methyl-4-4-nitrophenyl piperazine, 1-methyl-4-4-nitro-phenyl-piperazine, 4-methyl-1-4-nitrophenyl piperazine, acmc-1bvu1, cambridge id 5139345, 1-methyl-4-p-nitrophenyl piperazine, 1-4-nitrophenyl-4-methylpiperazine, 1-methyl-4-4-nitrophenyl piperizine, n-4-nitrophenyl-n'-methylpiperazine, 4-methyl-1-4-nitro-phenyl-piperazine
PubChem CID 2825198
IUPAC Name 1-methyl-4-(4-nitrophenyl)piperazine
SMILES CN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63665MD View Documents Alfa Aesar™
H63665MD
250mg Each for $28.84
 
AAH6366503 View Documents Alfa Aesar™
H6366503
1g Each for $85.14
 
AAH6366506 View Documents Alfa Aesar™
H6366506
5g Each for $337.73
 
Specifications

Chemical Identifiers

16155-03-6
221.26
GZNDUKANJZIZOT-UHFFFAOYSA-N
2825198
CN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
C11H15N3O2
MFCD00101003
1-methyl-4-4-nitrophenyl piperazine, 1-methyl-4-4-nitro-phenyl-piperazine, 4-methyl-1-4-nitrophenyl piperazine, acmc-1bvu1, cambridge id 5139345, 1-methyl-4-p-nitrophenyl piperazine, 1-4-nitrophenyl-4-methylpiperazine, 1-methyl-4-4-nitrophenyl piperizine, n-4-nitrophenyl-n'-methylpiperazine, 4-methyl-1-4-nitro-phenyl-piperazine
1-methyl-4-(4-nitrophenyl)piperazine

Specifications

250mg
C11H15N3O2
1-methyl-4-4-nitrophenyl piperazine, 1-methyl-4-4-nitro-phenyl-piperazine, 4-methyl-1-4-nitrophenyl piperazine, acmc-1bvu1, cambridge id 5139345, 1-methyl-4-p-nitrophenyl piperazine, 1-4-nitrophenyl-4-methylpiperazine, 1-methyl-4-4-nitrophenyl piperizine, n-4-nitrophenyl-n'-methylpiperazine, 4-methyl-1-4-nitro-phenyl-piperazine
CN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
221.26
221.26
97%
16155-03-6
MFCD00101003
GZNDUKANJZIZOT-UHFFFAOYSA-N
1-methyl-4-(4-nitrophenyl)piperazine
2825198
Solid
1-Methyl-4-(4-nitrophenyl)piperazine