missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-4-(4-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H6366506

 View more versions of this product

Catalog No. AAH6366506



Specifications

Specifications

1-Methyl-4-(4-nitrophenyl)piperazine
16155-03-6
MFCD00101003
GZNDUKANJZIZOT-UHFFFAOYSA-N
1-methyl-4-(4-nitrophenyl)piperazine
2825198
Solid
5g
C11H15N3O2
1-methyl-4-4-nitrophenyl piperazine, 1-methyl-4-4-nitro-phenyl-piperazine, 4-methyl-1-4-nitrophenyl piperazine, acmc-1bvu1, cambridge id 5139345, 1-methyl-4-p-nitrophenyl piperazine, 1-4-nitrophenyl-4-methylpiperazine, 1-methyl-4-4-nitrophenyl piperizine, n-4-nitrophenyl-n'-methylpiperazine, 4-methyl-1-4-nitro-phenyl-piperazine
CN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
221.26
221.26
97%

Chemical Identifiers

16155-03-6
221.26
GZNDUKANJZIZOT-UHFFFAOYSA-N
2825198
CN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
C11H15N3O2
MFCD00101003
1-methyl-4-4-nitrophenyl piperazine, 1-methyl-4-4-nitro-phenyl-piperazine, 4-methyl-1-4-nitrophenyl piperazine, acmc-1bvu1, cambridge id 5139345, 1-methyl-4-p-nitrophenyl piperazine, 1-4-nitrophenyl-4-methylpiperazine, 1-methyl-4-4-nitrophenyl piperizine, n-4-nitrophenyl-n'-methylpiperazine, 4-methyl-1-4-nitro-phenyl-piperazine
1-methyl-4-(4-nitrophenyl)piperazine
Documents