Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-4-(2-nitrophenyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-Methyl-4-(2-nitrophenyl)piperazine
CAS 62208-63-3
Formula Weight 221.26
MDL Number MFCD00156356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6336803

 
alfa aesar™
H6336803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6336806

 
alfa aesar™
H6336806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63368MD

 
alfa aesar™
H63368MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.26
Chemical Name or Material 1-Methyl-4-(2-nitrophenyl)piperazine
Physical Form Solid
Assay 97%
MDL Number MFCD00156356
CAS 62208-63-3
Quantity 1g