missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-4-(2-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H6336806

 View more versions of this product

Catalog No. AAH6336806



Specifications

Specifications

1-Methyl-4-(2-nitrophenyl)piperazine
62208-63-3
MFCD00156356
SSRLRVMFEGGGMQ-UHFFFAOYSA-N
1-methyl-4-(2-nitrophenyl)piperazine
410640
Solid
5g
C11H15N3O2
1-methyl-4-2-nitrophenyl piperazine, piperazine, 1-methyl-4-2-nitrophenyl, cbmicro_037654, 1-methyl-4-2-nitro-phenyl-piperazine, 2-4-methylpiperazin-1-yl nitrobenzene, 2-4-methylpiperazin-1-yl-1-nitrobenzene
CN1CCN(CC1)C2=CC=CC=C2[N+](=O)[O-]
221.26
221.26
97%

Chemical Identifiers

62208-63-3
221.26
SSRLRVMFEGGGMQ-UHFFFAOYSA-N
410640
CN1CCN(CC1)C2=CC=CC=C2[N+](=O)[O-]
C11H15N3O2
MFCD00156356
1-methyl-4-2-nitrophenyl piperazine, piperazine, 1-methyl-4-2-nitrophenyl, cbmicro_037654, 1-methyl-4-2-nitro-phenyl-piperazine, 2-4-methylpiperazin-1-yl nitrobenzene, 2-4-methylpiperazin-1-yl-1-nitrobenzene
1-methyl-4-(2-nitrophenyl)piperazine
SDS Documents