Please login to your online account to display your discounted pricing

1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride, 97+%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  AW00087DE

 View more versions of this product

Catalog No. AW00087DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 248.61
Quantity 5g
Assay 97%
Packaging Amber Glass Bottle
Molecular Formula C5H4ClF3N2O2S
CAS 519056-67-8
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H4ClF3N2O2S
Formula Weight 248.61