missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride, 97+%, Thermo Scientific™
Chemical Identifiers
CAS |
519056-67-8 |
Molecular Formula |
C5H4ClF3N2O2S |
Molecular Weight (g/mol) |
248.604 |
InChI Key |
HJFWSNIHZZKYIK-UHFFFAOYSA-N |
Synonym |
1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonylchloride,1-methyl-3-trifluoromethyl, chloro 1-methyl-3-trifluoromethyl pyrazol-4-yl sulfone, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonylchloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl |
PubChem CID |
2794561 |
IUPAC Name |
1-methyl-3-(trifluoromethyl)pyrazole-4-sulfonyl chloride |
SMILES |
CN1C=C(C(=N1)C(F)(F)F)S(=O)(=O)Cl |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
519056-67-8 |
|
248.604 |
|
1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonylchloride,1-methyl-3-trifluoromethyl, chloro 1-methyl-3-trifluoromethyl pyrazol-4-yl sulfone, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonylchloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl |
|
1-methyl-3-(trifluoromethyl)pyrazole-4-sulfonyl chloride |
|
C5H4ClF3N2O2S |
|
HJFWSNIHZZKYIK-UHFFFAOYSA-N |
|
2794561 |
|
CN1C=C(C(=N1)C(F)(F)F)S(=O)(=O)Cl |
Specifications
|
519056-67-8 |
|
Amber Glass Bottle |
|
1g |
|
HJFWSNIHZZKYIK-UHFFFAOYSA-N |
|
1-methyl-3-(trifluoromethyl)pyrazole-4-sulfonyl chloride |
|
2794561 |
|
97% |
|
97% |
|
C5H4ClF3N2O2S |
|
1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonylchloride,1-methyl-3-trifluoromethyl, chloro 1-methyl-3-trifluoromethyl pyrazol-4-yl sulfone, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonylchloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl |
|
CN1C=C(C(=N1)C(F)(F)F)S(=O)(=O)Cl |
|
248.604 |
|
248.61 |
|
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride |
SDS
missing translation for 'documents'