missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1-Methyl-3-thien-2-yl-1H-pyrazol-5-yl)methanol, 97%, Maybridge

Chemical Identifiers

CAS 879896-49-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H10N2OS
Formula Weight 194.26
MDL Number MFCD08435885
Chemical Name or Material (1-Methyl-3-thien-2-yl-1H-pyrazol-5-yl)methanol
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC47209DA SDS Fisher Scientific Company
CC47209DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

879896-49-8
100.0
C9H10N2OS
MFCD08435885
97%
96.5
97%
194.26
(1-Methyl-3-thien-2-yl-1H-pyrazol-5-yl)methanol

Specifications

194.26
100.0
97%
MFCD08435885
(1-Methyl-3-thien-2-yl-1H-pyrazol-5-yl)methanol
1g
879896-49-8
C9H10N2OS
96.5
97%