missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-3-propylimidazolium Tetrafluoroborate, TCI America™

$84.03 - $252.09

Chemical Identifiers

CAS 244193-48-4
Molecular Formula C7H13BF4N2
Molecular Weight (g/mol) 211.999
MDL Number MFCD14156016
InChI Key XFTQQJUVYZPNRC-UHFFFAOYSA-N
PubChem CID 11171853
IUPAC Name 1-methyl-3-propylimidazol-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CCCN1C=C[N+](=C1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M30365G View Documents TCI America
M30365G
5g Each for $84.03
 
M303625G View Documents TCI America
M303625G
25g Each for $252.09
 
Specifications

Chemical Identifiers

244193-48-4
211.999
XFTQQJUVYZPNRC-UHFFFAOYSA-N
1-methyl-3-propylimidazol-1-ium;tetrafluoroborate
C7H13BF4N2
MFCD14156016
11171853
[B-](F)(F)(F)F.CCCN1C=C[N+](=C1)C

Specifications

Yellow
5g
C7H13BF4N2
1760
[B-](F)(F)(F)F.CCCN1C=C[N+](=C1)C
211.999
212.00
≥98.0% (HPLC,N)
17°C
244193-48-4
MFCD14156016
XFTQQJUVYZPNRC-UHFFFAOYSA-N
1-methyl-3-propylimidazol-1-ium;tetrafluoroborate
11171853
Clear Liquid at 20°C
1-Methyl-3-propylimidazolium Tetrafluoroborate
Safety and Handling

Safety and Handling

TSCA : No