missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-3-propylimidazolium Bromide, TCI America™

$87.08 - $273.48

Chemical Identifiers

CAS 85100-76-1
Molecular Formula C7H13BrN2
MDL Number MFCD17014727
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M303425G View Documents TCI America
M303425G
25g Each for $273.48
 
M30345G View Documents TCI America
M30345G
5g Each for $87.08
 
Specifications

Chemical Identifiers

85100-76-1
C7H13BrN2
MFCD17014727

Specifications

Undesignated
85100-76-1
MFCD17014727
Clear Liquid at 20°C
1-Methyl-3-propylimidazolium Bromide
25g
C7H13BrN2
205.10
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No