missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-3-piperidinemethanol, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC163842500

 View more versions of this product

Catalog No. AC163842500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Methyl-3-piperidinemethanol
94°C
1.4772 to 1.4792
140.0°C to 145.0°C
7583-53-1
100.0
C7H15NO
1-methyl-3-hydroxymethylpiperidine
CN1CCCC(C1)CO
129.2
129.2
1.0100g/mL
Glass Bottle
1.01
250g
94.0
94% min. (GC)
21,II,8
UGXQXVDTGJCQHR-UHFFFAOYSA-N
(1-methylpiperidin-3-yl)methanol
97998
95%

Chemical Identifiers

7583-53-1
129.2
1-methyl-3-hydroxymethylpiperidine
(1-methylpiperidin-3-yl)methanol
C7H15NO
UGXQXVDTGJCQHR-UHFFFAOYSA-N
97998
CN1CCCC(C1)CO
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 231-488-5

SDS Documents