missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-3-piperidinemethanol, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC163840500

 View more versions of this product

Catalog No. AC163840500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Methyl-3-piperidinemethanol
94°C
1.4772 to 1.4792
50g
94.0
95%
1-methyl-3-piperidinemethanol, 1-methylpiperidin-3-yl methanol, 1-methylpiperidine-3-methanol, 3-piperidinemethanol, 1-methyl, 3-hydroxymethyl-1-methylpiperidine, 1-methyl-3-piperidyl methanol, 1-methyl-3-hydroxymethylpiperidine, 1-methyl-piperidin-3-yl-methanol, 3-hydroxymethyl-1-methylpiperidine, 3-piperidinemethanol,1-methyl-,--9ci
CN1CCCC(C1)CO
129.203
129.2
1.0100g/mL
Glass bottle
140.0°C to 145.0°C
7583-53-1
100.0
C7H15NO
UGXQXVDTGJCQHR-UHFFFAOYSA-N
(1-methylpiperidin-3-yl)methanol
97998
95%

Chemical Identifiers

7583-53-1
129.203
1-methyl-3-piperidinemethanol, 1-methylpiperidin-3-yl methanol, 1-methylpiperidine-3-methanol, 3-piperidinemethanol, 1-methyl, 3-hydroxymethyl-1-methylpiperidine, 1-methyl-3-piperidyl methanol, 1-methyl-3-hydroxymethylpiperidine, 1-methyl-piperidin-3-yl-methanol, 3-hydroxymethyl-1-methylpiperidine, 3-piperidinemethanol,1-methyl-,--9ci
(1-methylpiperidin-3-yl)methanol
C7H15NO
UGXQXVDTGJCQHR-UHFFFAOYSA-N
97998
CN1CCCC(C1)CO
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents