Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-3-phenylhydantoin, 95%

Chemical Identifiers

Chemical Name or Material 1-Methyl-3-phenylhydantoin
Formula Weight 190.2
MDL Number MFCD00464654
Synonym 1-Methyl-3-phenyl-2,4-imidazolidinedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6322003

 
alfa aesar™
H6322003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63220MD

 
alfa aesar™
H63220MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6322006

 
alfa aesar™
H6322006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.2
Chemical Name or Material 1-Methyl-3-phenylhydantoin
Physical Form Solid
Assay 95%
MDL Number MFCD00464654
Synonym 1-Methyl-3-phenyl-2,4-imidazolidinedione
Quantity 1g