missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-3-phenylhydantoin, 95%

Manufacturer:  Alfa Aesar™ H6322006

 View more versions of this product

Catalog No. AAH6322006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Methyl-3-phenylhydantoin
2221-12-7
MFCD00464654
YLKKRCYFGKGCKZ-UHFFFAOYSA-N
1-methyl-3-phenylimidazolidine-2,4-dione
10774030
Solid
5g
C10H10N2O2
1-methyl-3-phenylhydantoin, 2,4-imidazolidinedione, 1-methyl-3-phenyl
CN1CC(=O)N(C1=O)C2=CC=CC=C2
190.20
190.2
95%

Chemical Identifiers

2221-12-7
190.20
YLKKRCYFGKGCKZ-UHFFFAOYSA-N
10774030
CN1CC(=O)N(C1=O)C2=CC=CC=C2
C10H10N2O2
MFCD00464654
1-methyl-3-phenylhydantoin, 2,4-imidazolidinedione, 1-methyl-3-phenyl
1-methyl-3-phenylimidazolidine-2,4-dione
Documents