Please login to your online account to display your discounted pricing

1-Methyl-3-phenyl-1H-pyrazole-5-carbonyl chloride, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC46602DA

 View more versions of this product

Catalog No. CC46602DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 220.66
Quantity 1g
Assay 95%
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C11H9ClN2O
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violently with water.
CAS Min % 94.5
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD08271940
CAS 864068-95-1
Chemical Name or Material 1-Methyl-3-phenyl-1H-pyrazole-5-carbonyl chloride

Specifications

Chemical Name or Material 1-Methyl-3-phenyl-1H-pyrazole-5-carbonyl chloride
CAS 864068-95-1
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H9ClN2O
Formula Weight 220.66
MDL Number MFCD08271940
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents