missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-3-phenyl-1H-pyrazole-5-carbonyl chloride, 95%, Maybridge

Chemical Identifiers

CAS 864068-95-1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C11H9ClN2O
Formula Weight 220.66
MDL Number MFCD08271940
Chemical Name or Material 1-Methyl-3-phenyl-1H-pyrazole-5-carbonyl chloride
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC46602DA SDS Fisher Scientific Company
CC46602DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC46602CB SDS Fisher Scientific Company
CC46602CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

864068-95-1
100.0
C11H9ClN2O
MFCD08271940
95%
94.5
95%
220.66
1-Methyl-3-phenyl-1H-pyrazole-5-carbonyl chloride

Specifications

220.66
100.0
95%
Amber Glass Bottle
864068-95-1
95%
1g
C11H9ClN2O
94.5
MFCD08271940
1-Methyl-3-phenyl-1H-pyrazole-5-carbonyl chloride

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger